accessibility menu, dialog, popup

UserName

BNClClHOOH

CAS RN 951677-39-7

IUPAC Name: (2,3-dichloropyridin-4-yl)boronic acid
Synonyms: 2,3-dichloropyridin-4-ylboronic acid
Molecular Weight (g/mol): 191.8
Molecular Formula: C5H4BCl2NO2
InChi Key: ABRTZUPJNVJMHP-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results