missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropropanoic Acid, TRC

Product Code. 30381091
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381091 1 g -
30380674 100 mg -
30382927 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381091 Supplier Toronto Research Chemicals Supplier No. TRCD4346301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-64-0
Molecular Formula C3 H4 Cl2 O2
Molecular Weight (g/mol) 142.96
Synonym Propionic acid, 2,3-dichloro- (8CI), 2,3-Dichloropropanoic acid, 2,3-Dichloropropionic acid, NSC 60521, α,β-Dichloropropionic acid, 2,3-Dichloropropionic Acid
IUPAC Name 2,3-dichloropropanoic acid
SMILES OC(=O)C(Cl)CCl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H4Cl2O2/c4-1-2(5)3(6)7/h2H,1H2,(H,6,7)
Formula Weight 141.9588
Chemical Name or Material 2,3-Dichloropropanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.