CAS RN 565-64-0
IUPAC Name:
2,3-dichloropropanoic acid
Synonyms:
2,3-dichloropropionic acid
Molecular Weight (g/mol):
142.96
Molecular Formula:
C3H4Cl2O2
InChi Key:
GKFWNPPZHDYVLI-UHFFFAOYNA-N
SMILES:
OC(=O)C(Cl)CCl
1
–
1
of
1
results
1
2,3-Dichloropropanoic Acid, TRC
CAS: 565-64-0 Molecular Formula: C3 H4 Cl2 O2 Molecular Weight (g/mol): 142.96 Synonym: Propionic acid, 2,3-dichloro- (8CI),2,3-Dichloropropanoic acid,2,3-Dichloropropionic acid,NSC 60521,α,β-Dichloropropionic acid,2,3-Dichloropropionic Acid IUPAC Name: 2,3-dichloropropanoic acid SMILES: OC(=O)C(Cl)CCl