missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diamino-4-chloropyridine, TRC

Product Code. 30357548
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357548 250 mg 250mg
30365968 100 mg 100mg
30367910 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357548 Supplier Toronto Research Chemicals Supplier No. TRCD415090250MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24484-98-8
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.57
IUPAC Name 4-chloropyridine-2,3-diamine
SMILES Nc1nccc(Cl)c1N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H6ClN3/c6-3-1-2-9-5(8)4(3)7/h1-2H,7H2,(H2,8,9)
Formula Weight 143.025
Chemical Name or Material 2,3-Diamino-4-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.