CAS RN 24484-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,3-Diamino-4-chloropyridine, TRC
CAS: 24484-98-8 Molecular Formula: C5H6ClN3 Molecular Weight (g/mol): 143.57 IUPAC Name: 4-chloropyridine-2,3-diamine SMILES: Nc1nccc(Cl)c1N