missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trichloropyridine, ≥98%, Thermo Scientific Chemicals
£54.80 - £246.00
Chemical Identifiers
CAS | 16063-70-0 |
---|---|
Molecular Formula | C5H2Cl3N |
Molecular Weight (g/mol) | 182.428 |
MDL Number | MFCD00043007 |
InChI Key | CNLIIAKAAMFCJG-UHFFFAOYSA-N |
Synonym | pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005 |
PubChem CID | 27666 |
IUPAC Name | 2,3,5-trichloropyridine |
SMILES | C1=C(C=NC(=C1Cl)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15451259
|
Thermo Scientific Alfa Aesar
H64173.22 |
100 g |
£54.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15471259
|
Thermo Scientific Alfa Aesar
H64173.A1 |
1 kg | |||||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Chemical Identifiers
16063-70-0 | |
182.428 | |
CNLIIAKAAMFCJG-UHFFFAOYSA-N | |
27666 | |
C1=C(C=NC(=C1Cl)Cl)Cl |
C5H2Cl3N | |
MFCD00043007 | |
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005 | |
2,3,5-trichloropyridine |
Specifications
16063-70-0 | |
White to Yellow | |
113°C (235°F) | |
MFCD00043007 | |
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005 | |
CNLIIAKAAMFCJG-UHFFFAOYSA-N | |
2,3,5-trichloropyridine | |
27666 | |
≥98% | |
2,3,5-Trichloropyridine |
46°C to 50°C (literature) | |
219°C (literature) | |
C5H2Cl3N | |
119384 | |
Slightly soluble in water. | |
C1=C(C=NC(=C1Cl)Cl)Cl | |
182.428 | |
182.43 | |
Solid |
Safety and Handling
missing translation for 'einecsNumber' : 407-270-2
missing translation for 'rtecsNumber' : UU0525000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only