missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trichloropyridine, 98%
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
It is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Specifications
| Melting Point | 46°C to 50°C (literature) |
| Color | White to Yellow |
| Boiling Point | 219°C (literature) |
| Flash Point | 113°C (235°F) |
| Quantity | 100 g |
| Beilstein | 119384 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 182.43 |
| Percent Purity | ≥98% |
| Physical Form | Solid |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction