missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-difluorophenyl)acetic acid, Thermo Scientific™

Catalog Number 10066992
Click to view available options
Quantity:
10 g
100 g
This item is not returnable. View return policy
105184-38-1
C8H6F2O2
172.131
IGGNSAVLXJKCNH-UHFFFAOYSA-N
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
145424
2-(3,5-difluorophenyl)acetic acid
C1=C(C=C(C=C1F)F)CC(=O)O
This item is not returnable. View return policy
105184-38-1
C8H6F2O2
172.131
IGGNSAVLXJKCNH-UHFFFAOYSA-N
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
145424
2-(3,5-difluorophenyl)acetic acid
C1=C(C=C(C=C1F)F)CC(=O)O
Assay Percent Range 97%
Quantity 10 g
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 2-(3, 5-Difluorophenyl)acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'