missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoropyridine-4-acetic acid, 98%, Thermo Scientific™

Product Code. 15436647
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15436647 1 g 1g
15418645 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15436647

Brand: Thermo Scientific Alfa Aesar H33785.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65697-73-6
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD02944320
InChI Key SOHLFVREHOCIMD-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-pyridinyl acetic acid, 2,3,5,6-tetrafluoropyridine-4-yl acetic acid, 2-perfluoropyridin-4-yl acetic acid, 2,3,5,6-tetrafluoropyridine-4-acetic acid, 2,3,5,6-tetrafluoropyridin-4-yl acetic acid, tetrafluoropyridin-4-yl acetic acid, 2-2,3,5,6-tetrafluoropyridin-4-yl acetic acid
PubChem CID 1237119
IUPAC Name 2-(2,3,5,6-tetrafluoropyridin-4-yl)acetic acid
SMILES C(C1=C(C(=NC(=C1F)F)F)F)C(=O)O

Specifications

Melting Point 145°C to 149°C
Quantity 1 g
Formula Weight 209.1
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluoropyridine-4-acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.