accessibility menu, dialog, popup

UserName

CAS RN 65697-73-6

NFFFFOOH

CAS RN 65697-73-6

IUPAC Name: 2-(2,3,5,6-tetrafluoropyridin-4-yl)acetic acid
Synonyms: (2,3,5,6-tetrafluoropyridin-4-yl)acetic acid
Molecular Weight (g/mol): 209.1
Molecular Formula: C7H3F4NO2
InChi Key: SOHLFVREHOCIMD-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C(F)=NC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results