missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluoroanisole, 98%

Product Code. 11329829
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329829 100 g 100g
11309829 5 g 5g
11319829 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329829 Supplier Thermo Scientific Alfa Aesar Supplier No. B23928.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 389-40-2 | C7H3F5O | 198.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389-40-2
Molecular Formula C7H3F5O
Molecular Weight (g/mol) 198.092
MDL Number MFCD00000289
InChI Key ZRQUIRABLIQJRI-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluoroanisole, pentafluoroanisole, benzene, pentafluoromethoxy, pentafluorophenyl methyl ether, pentafluoromethoxybenzene, methyl pentafluorophenyl ether, benzene, 1,2,3,4,5-pentafluoro-6-methoxy, anisole, 2,3,4,5,6-pentafluoro, 1,2,3,4,6-pentafluoro-5-methoxybenzene, methoxypentafluorobenzene
PubChem CID 67849
IUPAC Name 1,2,3,4,5-pentafluoro-6-methoxybenzene
SMILES COC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point -38°C
Density 1.5
Boiling Point 136°C to 138°C
Flash Point 32°C (89°F)
Refractive Index 1.409
Quantity 100 g
UN Number UN1993
Beilstein 2052664
Formula Weight 198.09
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluoroanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.