accessibility menu, dialog, popup

UserName

CAS RN 389-40-2

IUPAC Name: 1,2,3,4,5-pentafluoro-6-methoxybenzene
Synonyms: benzene, pentafluoromethoxy-
Molecular Weight (g/mol): 198.09
Molecular Formula: C7H3F5O
InChi Key: ZRQUIRABLIQJRI-UHFFFAOYSA-N
SMILES: COC1=C(F)C(F)=C(F)C(F)=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results