missing translation for 'onlineSavingsMsg'
Learn More

2,3,3,5-Tetramethylindolenine, 94%

Product Code. 15465897
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15465897

Brand: Thermo Scientific Alfa Aesar H33109.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25981-82-2 | C12H15N | 173.26 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25981-82-2
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD03093020
InChI Key RQVAPBRSUHSDGP-UHFFFAOYSA-N
Synonym 2,3,3,5-tetramethyl-3h-indole, 2,3,3,5-tetramethylindolenine, 3h-indole, 2,3,3,5-tetramethyl, pubchem2092, 2,3,3,5-tetramethyl-3h-indol, 2,3,3,5-tetramethyl-3-h-indole, 3h-indole,2,3,3,5-tetramethyl, 2,3,3,5-tetramethyl-3-hydroindole, 2,3,3,5-tetramethyl-3-h indole
PubChem CID 117693
IUPAC Name 2,3,3,5-tetramethylindole
SMILES CC1=NC2=CC=C(C)C=C2C1(C)C

Specifications

Quantity 10 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 173.26
Percent Purity 96%
Chemical Name or Material 2,3,3,5-Tetramethylindolenine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.