CAS RN 25981-82-2
CAS RN 25981-82-2
IUPAC Name:
2,3,3,5-tetramethyl-3H-indole
Synonyms:
2,3,3,5-tetramethylindole
Molecular Weight (g/mol):
173.26
Molecular Formula:
C12H15N
InChi Key:
RQVAPBRSUHSDGP-UHFFFAOYSA-N
SMILES:
CC1=NC2=CC=C(C)C=C2C1(C)C
1
–
1
of
1
results
1
2,3,3,5-Tetramethylindolenine, 94%
CAS: 25981-82-2 Molecular Formula: C12H15N Molecular Weight (g/mol): 173.26 MDL Number: MFCD03093020 InChI Key: RQVAPBRSUHSDGP-UHFFFAOYSA-N Synonym: 2,3,3,5-tetramethyl-3h-indole,2,3,3,5-tetramethylindolenine,3h-indole, 2,3,3,5-tetramethyl,pubchem2092,2,3,3,5-tetramethyl-3h-indol,2,3,3,5-tetramethyl-3-h-indole,3h-indole,2,3,3,5-tetramethyl,2,3,3,5-tetramethyl-3-hydroindole,2,3,3,5-tetramethyl-3-h indole PubChem CID: 117693 IUPAC Name: 2,3,3,5-tetramethylindole SMILES: CC1=NC2=CC=C(C)C=C2C1(C)C