missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic Acid, TRC

Product Code. 30399106
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399106 5 g
30400289 500 mg
30401132 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399106 Supplier Toronto Research Chemicals Supplier No. TRCF5919605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 391211-97-5
Molecular Formula C13H7F3INO2
Molecular Weight (g/mol) 393.1
Synonym 3,4-Difluoro-2-[(2-fluoro-4-iodophenyl)amino]benzoic Acid, 3,4-Difluoro-2-(2-fluoro-4-iodophenylamino)benzoic Acid
SMILES OC(=O)c1ccc(F)c(F)c1Nc2ccc(I)cc2F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H7F3INO2/c14-8-3-2-7(13(19)20)12(11(8)16)18-10-4-1-6(17)5-9(10)15/h1-5,18H,(H,19,20)
Chemical Name or Material 2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.