CAS RN 391211-97-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic Acid, TRC
CAS: 391211-97-5 Molecular Formula: C13H7F3INO2 Molecular Weight (g/mol): 393.1 Synonym: 3,4-Difluoro-2-[(2-fluoro-4-iodophenyl)amino]benzoic Acid,3,4-Difluoro-2-(2-fluoro-4-iodophenylamino)benzoic Acid SMILES: OC(=O)c1ccc(F)c(F)c1Nc2ccc(I)cc2F