missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethylcyclohexanone, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -20.0°C |
Color | Colorless to Yellow |
Density | 0.9120g/mL |
Boiling Point | 169.0°C to 170.0°C (0.8 mmHg) |
Flash Point | 50°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | (CH3)2C6H8(=O) |
Refractive Index | 1.4470 to 1.4490 (20°C, 589nm) |
Quantity | 5 g |
Show More |
Safety and Handling
Product Identifier
- 2,2-Dimethylcyclohexanone
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'