CAS RN 1193-47-1
IUPAC Name:
2,2-dimethylcyclohexan-1-one
Synonyms:
cyclohexanone, 2,2-dimethyl-
Molecular Weight (g/mol):
126.2
Molecular Formula:
C8H14O
InChi Key:
KNSPBSQWRKKAPI-UHFFFAOYSA-N
SMILES:
CC1(C)CCCCC1=O
1
–
1
of
1
results
1
2,2-Dimethylcyclohexanone
CAS: 1193-47-1 Molecular Formula: C8H14O Molecular Weight (g/mol): 126.2 InChI Key: KNSPBSQWRKKAPI-UHFFFAOYSA-N Synonym: 2,2-dimethylcyclohexanone,cyclohexanone, 2,2-dimethyl,dimethylcyclohexanone,cyclohexanone, dimethyl,6,6-dimethylcyclohexanone,cyclohexanone,2-dimethyl,acmc-20a0u6,2,2-dimethyl-cyclohexanone,cyclohexanone,2,2-dimethyl,2,2-dimethyl-1-cyclohexanone PubChem CID: 136929 IUPAC Name: 2,2-dimethylcyclohexan-1-one SMILES: CC1(CCCCC1=O)C