missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydrobenzofuran-3,7-diol, TRC

Product Code. 30380116
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380116 250 mg
30376104 50 mg
30383200 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380116 Supplier Toronto Research Chemicals Supplier No. TRCD482241250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17781-15-6
Molecular Formula C10 H12 O3
Molecular Weight (g/mol) 180.2
Synonym 2,3-Dihydro-2,2-dimethyl-3,7-benzofurandiol, 2,2-Dimethyl-2,3-dihydro-1-benzofuran-3,7-diol, 2,2-Dimethyl-3,7-dihydroxy-2,3-dihydrobenzofuran, 2,3-Dihydro-2,2-dimethyl-3,7-dihydroxybenzofuran, 2,3-Dihydro-3,7-dihydroxy-2,2-dimethylbenzofuran, 3,7-Dihydroxy-2,2-dimethyl-2,3-dihydrobenzofura
IUPAC Name 2,2-dimethyl-3H-1-benzofuran-3,7-diol
SMILES CC1(C)Oc2c(O)cccc2C1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H12O3/c1-10(2)9(12)6-4-3-5-7(11)8(6)13-10/h3-5,9,11-12H,1-2H3
Formula Weight 180.0786
Chemical Name or Material Carbofuran-3-hydroxy-7-phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.