CAS RN 17781-15-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2-Dimethyl-2,3-dihydrobenzofuran-3,7-diol, TRC
CAS: 17781-15-6 Molecular Formula: C10 H12 O3 Molecular Weight (g/mol): 180.2 Synonym: 2,3-Dihydro-2,2-dimethyl-3,7-benzofurandiol,2,2-Dimethyl-2,3-dihydro-1-benzofuran-3,7-diol,2,2-Dimethyl-3,7-dihydroxy-2,3-dihydrobenzofuran,2,3-Dihydro-2,2-dimethyl-3,7-dihydroxybenzofuran,2,3-Dihydro-3,7-dihydroxy-2,2-dimethylbenzofuran,3,7-Dihydroxy-2,2-dimethyl-2,3-dihydrobenzofura IUPAC Name: 2,2-dimethyl-3H-1-benzofuran-3,7-diol SMILES: CC1(C)Oc2c(O)cccc2C1O