missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorocyclopropanemethanol, 97%

Product Code. 15494757
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494757 250 mg 250mg
15484757 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494757 Supplier Thermo Scientific Alfa Aesar Supplier No. H32083.MD
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 509072-57-5 | C4H6F2O | 108.088 g/mol

2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 509072-57-5
Molecular Formula C4H6F2O
Molecular Weight (g/mol) 108.088
MDL Number MFCD06797748
InChI Key XOLSMTBBIZDHSG-UHFFFAOYSA-N
Synonym 2,2-difluorocyclopropyl methanol, 2,2-difluorocyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 97+%, soqdfiarbswxuup@, 2,2-difluoro-cyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 2,2-difluoro-cyclopropyl-methanol, cyclopropanemethanol, 2,2-difluoro
PubChem CID 2761401
IUPAC Name (2,2-difluorocyclopropyl)methanol
SMILES C1C(C1(F)F)CO

Specifications

Boiling Point 129°C
Quantity 250 mg
UN Number UN1987
Formula Weight 108.09
Percent Purity 97%
Chemical Name or Material 2,2-Difluorocyclopropanemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.