CAS RN 509072-57-5
CAS RN 509072-57-5
IUPAC Name:
(2,2-difluorocyclopropyl)methanol
Synonyms:
(2,2-difluorocyclopropyl)methanol
Molecular Weight (g/mol):
108.09
Molecular Formula:
C4H6F2O
InChi Key:
XOLSMTBBIZDHSG-UHFFFAOYNA-N
SMILES:
OCC1CC1(F)F
1
–
1
of
1
results
1
2,2-Difluorocyclopropanemethanol, 97%
CAS: 509072-57-5 Molecular Formula: C4H6F2O Molecular Weight (g/mol): 108.088 MDL Number: MFCD06797748 InChI Key: XOLSMTBBIZDHSG-UHFFFAOYSA-N Synonym: 2,2-difluorocyclopropyl methanol,2,2-difluorocyclopropanemethanol,2,2-difluorocyclopropyl-methanol, 97+%,soqdfiarbswxuup@,2,2-difluoro-cyclopropanemethanol,2,2-difluorocyclopropyl-methanol,2,2-difluoro-cyclopropyl-methanol,cyclopropanemethanol, 2,2-difluoro PubChem CID: 2761401 IUPAC Name: (2,2-difluorocyclopropyl)methanol SMILES: C1C(C1(F)F)CO