missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloro-1-(2,4-dichlorophenyl)ethanone, TRC

Product Code. 30387669
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387669 10 g
30371436 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387669 Supplier Toronto Research Chemicals Supplier No. TRCD43412510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2274-66-0
Molecular Formula C8H4Cl4O
Molecular Weight (g/mol) 257.93
Synonym 2,2,2',4'-Tetrachloroacetophenone, 2,4-Dichlorophenacylidene Dichloride, NSC 81222, alpha,alpha,2',4'-Tetrachloroacetophenone
IUPAC Name 2,2-dichloro-1-(2,4-dichlorophenyl)ethanone
SMILES ClC(Cl)C(=O)c1ccc(Cl)cc1Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H4Cl4O/c9-4-1-2-5(6(10)3-4)7(13)8(11)12/h1-3,8H
Formula Weight 255.9016
Chemical Name or Material 2,2-Dichloro-1-(2,4-dichlorophenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.