CAS RN 2274-66-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2-Dichloro-1-(2,4-dichlorophenyl)ethanone, TRC
CAS: 2274-66-0 Molecular Formula: C8H4Cl4O Molecular Weight (g/mol): 257.93 Synonym: 2,2,2',4'-Tetrachloroacetophenone,2,4-Dichlorophenacylidene Dichloride,NSC 81222,alpha,alpha,2',4'-Tetrachloroacetophenone IUPAC Name: 2,2-dichloro-1-(2,4-dichlorophenyl)ethanone SMILES: ClC(Cl)C(=O)c1ccc(Cl)cc1Cl