missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Diamino-2'-deoxyadenosine, 98%, Thermo Scientific Chemicals

Product Code. 15457059
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15457059 Supplier Thermo Scientific Alfa Aesar Supplier No. J65748.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 215943-79-6 | Molecular Formula: C10H15N7O3 | Molecular Weight: 281.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215943-79-6
Molecular Formula C10H15N7O3
Molecular Weight (g/mol) 281.28
MDL Number MFCD09750839
InChI Key DIFJLWYUVQLAHS-SJLGBTOWNA-N
Synonym 2,2'-diamino-2'-deoxyadenosine, 2'-amino-2'-deoxy-2,6-diaminopurineriboside, 2r,3s,4r,5r-4-amino-5-2,6-diamino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, 2r,3s,4r,5r-4-amino-5-2,6-diaminopurin-9-yl-2-hydroxymethyl oxolan-3-ol
PubChem CID 71744251
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=C(N)N=C12

Tekniske data

Color White
Quantity 1 g
IUPAC Name 4-amino-5-(2,6-diamino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 281.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,2'-Diamino-2'-deoxyadenosine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.