missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-butyl-1,3-propanediol, 96%, Thermo Scientific™

Product Code. 11378356
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11378356

Brand: Thermo Scientific Alfa Aesar L15619.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,2-Di-n-butyl-1,3-propanediol is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24765-57-9
Molecular Formula C11H24O2
Molecular Weight (g/mol) 188.311
MDL Number MFCD01863725
InChI Key OJMJOSRCBAXSAQ-UHFFFAOYSA-N
Synonym 2,2-di-n-butyl-1,3-propanediol, 2,2-dibutyl-1,3-propanediol, 5,5-nonanedimethanol, 2,2,-dibutylpropane-1,3-diol, 2,2-dibutyl-propane-1,3-diol, 2,2-di-n-butyl-1,3-propandiol
PubChem CID 4080430
IUPAC Name 2,2-dibutylpropane-1,3-diol
SMILES CCCCC(CCCC)(CO)CO

Specifications

Melting Point 38°C to 42°C
Boiling Point 125°C to 130°C (1 mmHg)
Quantity 25 g
Beilstein 1699746
Solubility Information Soluble in water.
Formula Weight 188.31
Percent Purity 96%
Chemical Name or Material 2,2-Di-n-butyl-1,3-propanediol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.