accessibility menu, dialog, popup

UserName

CAS RN 24765-57-9

CH3H3CHOHO

CAS RN 24765-57-9

IUPAC Name: 2,2-dibutylpropane-1,3-diol
Synonyms: 2,2-dibutylpropane-1,3-diol
Molecular Weight (g/mol): 188.31
Molecular Formula: C11H24O2
InChi Key: OJMJOSRCBAXSAQ-UHFFFAOYSA-N
SMILES: CCCCC(CO)(CO)CCCC
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results