missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Butoxyethoxy)acetic Acid, TRC

Product Code. 30240895
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240895 100 mg 100mg
30240782 50 mg 50mg
30240486 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240895 Supplier Toronto Research Chemicals Supplier No. TRCB415743100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82941-26-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
Synonym (2-Butoxyethoxy)acetic acid, (2-n-Butoxyethoxy)acetic acid, 3,6-Dioxadecanoic acid
IUPAC Name 2-(2-butoxyethoxy)acetic acid
SMILES CCCCOCCOCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H16O4/c1-2-3-4-11-5-6-12-7-8(9)10/h2-7H2,1H3,(H,9,10)
Formula Weight 176.1049
Chemical Name or Material 2-(2-Butoxyethoxy)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.