CAS RN 82941-26-2
CAS RN 82941-26-2
IUPAC Name:
2-butoxyethyl ethaneperoxoate
Synonyms:
2-butoxyethyl ethaneperoxoate
Molecular Weight (g/mol):
176.21
Molecular Formula:
C8H16O4
InChi Key:
OUINRTZUFNPIBX-UHFFFAOYSA-N
SMILES:
CCCCOCCOOC(C)=O
1
–
1
of
1
results
1
2-(2-Butoxyethoxy)acetic Acid, TRC
CAS: 82941-26-2 Molecular Formula: C8H16O4 Molecular Weight (g/mol): 176.21 Synonym: (2-Butoxyethoxy)acetic acid,(2-n-Butoxyethoxy)acetic acid,3,6-Dioxadecanoic acid IUPAC Name: 2-(2-butoxyethoxy)acetic acid SMILES: CCCCOCCOCC(=O)O