missing translation for 'onlineSavingsMsg'
Learn More

2-(2,5-Dimethoxyphenyl)-N-(2-methoxybenzyl)ethanamine Hydrochloride, TRC

Product Code. 30386358
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386358 10 mg
30371711 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386358 Supplier Toronto Research Chemicals Supplier No. TRCD47129010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1566571-52-5
Molecular Formula C18 H23 N O3 . HCl
Molecular Weight (g/mol) 337.841
Synonym 25H-NBOMe HCl, Benzeneethanamine, 2,5-dimethoxy-N-[(2-methoxyphenyl)methyl]-, hydrochloride (1:1), 2-(2,5-Dimethoxyphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine hydrochloride
IUPAC Name 2-(2,5-dimethoxyphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine;hydrochloride
SMILES Cl.COc1ccc(OC)c(CCNCc2ccccc2OC)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H23NO3.ClH/c1-20-16-8-9-18(22-3)14(12-16)10-11-19-13-15-6-4-5-7-17(15)21-2;/h4-9,12,19H,10-11,13H2,1-3H3;1H
Formula Weight 337.144
Chemical Name or Material 25H-NBOMe (hydrochloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.