CAS RN 1566571-52-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(2,5-Dimethoxyphenyl)-N-(2-methoxybenzyl)ethanamine Hydrochloride, TRC
CAS: 1566571-52-5 Molecular Formula: C18 H23 N O3 . HCl Molecular Weight (g/mol): 337.841 Synonym: 25H-NBOMe HCl,Benzeneethanamine, 2,5-dimethoxy-N-[(2-methoxyphenyl)methyl]-, hydrochloride (1:1),2-(2,5-Dimethoxyphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine hydrochloride IUPAC Name: 2-(2,5-dimethoxyphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine;hydrochloride SMILES: Cl.COc1ccc(OC)c(CCNCc2ccccc2OC)c1