missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluorocyclobutanol, 95%

Product Code. 11384056
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384056 5 g 5g
11374056 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384056 Supplier Thermo Scientific Alfa Aesar Supplier No. L19186.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 374-32-3 | C4H4F4O | 144.069 g/mol

2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-32-3
Molecular Formula C4H4F4O
Molecular Weight (g/mol) 144.069
MDL Number MFCD00190085
InChI Key FNENUTDUAXPVTF-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluorocyclobutanol, cyclobutanol, 2,2,3,3-tetrafluoro, cyclobutanol,2,2,3,3-tetrafluoro
PubChem CID 2776621
IUPAC Name 2,2,3,3-tetrafluorocyclobutan-1-ol
SMILES C1C(C(C1(F)F)(F)F)O

Specifications

Density 1.414
Boiling Point 155°C to 157°C
Flash Point 44°C (111°F)
Refractive Index 1.368
Quantity 5 g
UN Number UN1987
Beilstein 2429286
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 144.07
Percent Purity 95%
Chemical Name or Material 2,2,3,3-Tetrafluorocyclobutanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.