accessibility menu, dialog, popup

UserName

CAS RN 374-32-3

IUPAC Name: 2,2,3,3-tetrafluorocyclobutan-1-ol
Synonyms: 2,2,3,3-tetrafluorocyclobutan-1-ol
Molecular Weight (g/mol): 144.07
Molecular Formula: C4H4F4O
InChi Key: FNENUTDUAXPVTF-UHFFFAOYNA-N
SMILES: OC1CC(F)(F)C1(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results