missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene, 97%

Product Code. 11489737
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489737 10 g 10g
11499737 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489737 Supplier Thermo Scientific Alfa Aesar Supplier No. B22730.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1665-79-8 | C7H15O5P | 210.166 g/mol

2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-79-8
Molecular Formula C7H15O5P
Molecular Weight (g/mol) 210.166
MDL Number MFCD00014510
InChI Key FMHPXHSMBAZDOS-UHFFFAOYSA-N
Synonym 2,2,2-trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene, 1,3,2-dioxaphosphole, 2,2-dihydro-2,2,2-trimethoxy-4,5-dimethyl, 2,2,2-trimethoxy-4,5-dimethyl-1,3-dioxaphospholene, 2,2,2-trimethoxy-4,5-dimethyl-1,3,2-dioxaphosphole, 2,2,2-trimethoxy-4,5-dimethyl-1,3,2??-dioxaphosphole, acmc-20aokr, 2,2,2-trimethoxy-4,5-dimethyl-1,3,2, e5-dioxaphosphole, 2lambda5-1,3,2-dioxaphosphole, 2,2,2-trimethoxy-4,5-dimethyl, 2,2-dihydro-2,2,2-trimethoxy-4,5-dimethyl-1,3,2-dioxaphosphole
PubChem CID 74272
IUPAC Name 2,2,2-trimethoxy-4,5-dimethyl-1,3,2$l^{5}-dioxaphosphole
SMILES CC1=C(OP(O1)(OC)(OC)OC)C

Specifications

Boiling Point 64°C to 66°C (1 mmHg)
Quantity 10 g
Formula Weight 210.17
Percent Purity 97%
Chemical Name or Material 2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.