accessibility menu, dialog, popup

UserName

CH3OPOH3COH3COH3COCH3

CAS RN 1665-79-8

IUPAC Name: 2,2,2-trimethoxy-4,5-dimethyl-2H-1,3,2λ⁵-dioxaphosphole
Synonyms: 2,2,2-trimethoxy-4,5-dimethyl-1,3,2λ⁵-dioxaphosphole
Molecular Weight (g/mol): 210.17
Molecular Formula: C7H15O5P
InChi Key: FMHPXHSMBAZDOS-UHFFFAOYSA-N
SMILES: COP1(OC)(OC)OC(C)=C(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results