missing translation for 'onlineSavingsMsg'
Learn More

2-((1S,2S,5S)-6,6-Dimethylbicyclo[3.1.1]heptan-2-yl)ethanol, TRC

Product Code. 30381653
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381653 1 g 1g
30369031 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381653 Supplier Toronto Research Chemicals Supplier No. TRCD4648851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133001-09-9
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
Synonym (1S,2S,5S)-6,6-Dimethylbicyclo[3.1.1]heptane-2-ethanol, [1S-(1alpha,2beta,5alpha)]-6,6-Dimethylbicyclo[3.1.1]heptane-2-ethanol
IUPAC Name 2-[(1S,4R,5S)-6,6-dimethyl-4-bicyclo[3.1.1]heptanyl]ethanol
SMILES CC1(C)[C@H]2CC[C@H](CCO)[C@@H]1C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H20O/c1-11(2)9-4-3-8(5-6-12)10(11)7-9/h8-10,12H,3-7H2,1-2H3/t8-,9+,10+/m1/s1
Formula Weight 168.1514
Chemical Name or Material 2-((1S,2S,5S)-6,6-Dimethylbicyclo[3.1.1]heptan-2-yl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.