CAS RN 133001-09-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-((1S,2S,5S)-6,6-Dimethylbicyclo[3.1.1]heptan-2-yl)ethanol, TRC
CAS: 133001-09-9 Molecular Formula: C11H20O Molecular Weight (g/mol): 168.28 Synonym: (1S,2S,5S)-6,6-Dimethylbicyclo[3.1.1]heptane-2-ethanol,[1S-(1alpha,2beta,5alpha)]-6,6-Dimethylbicyclo[3.1.1]heptane-2-ethanol IUPAC Name: 2-[(1S,4R,5S)-6,6-dimethyl-4-bicyclo[3.1.1]heptanyl]ethanol SMILES: CC1(C)[C@H]2CC[C@H](CCO)[C@@H]1C2