missing translation for 'onlineSavingsMsg'
Learn More

2-[(1S,2S)-1-Ethyl-2-(phenylmethoxy)propyl]-2,4-dihydro-4-[4-[4-(4-hydroxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one, TRC

Product Code. 30380553
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380553 250 mg 250mg
30374265 100 mg -
30372264 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380553 Supplier Toronto Research Chemicals Supplier No. TRCE925535250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184177-83-1
Molecular Formula C30 H35 N5 O3
Molecular Weight (g/mol) 513.63
Synonym [S-(R*,R*)]-2-[1-Ethyl-2-(phenylmethoxy)propyl]-2,4-dihydro-4-[4-[4-(4-hydroxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one, Posaconazole Impurity
IUPAC Name 4-[4-[4-(4-hydroxyphenyl)piperazin-1-yl]phenyl]-2-[(2S,3S)-2-phenylmethoxypentan-3-yl]-1,2,4-triazol-3-one
SMILES CC[C@@H]([C@H](C)OCc1ccccc1)N2N=CN(C2=O)c3ccc(cc3)N4CCN(CC4)c5ccc(O)cc5

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C30H35N5O3/c1-3-29(23(2)38-21-24-7-5-4-6-8-24)35-30(37)34(22-31-35)27-11-9-25(10-12-27)32-17-19-33(20-18-32)26-13-15-28(36)16-14-26/h4-16,22-23,29,36H,3,17-21H2,1-2H3/t23-,29-/m0/s1
Formula Weight 513.274
Chemical Name or Material 2-[(1S,2S)-1-Ethyl-2-(phenylmethoxy)propyl]-2,4-dihydro-4-[4-[4-(4-hydroxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.