CAS RN 184177-83-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[(1S,2S)-1-Ethyl-2-(phenylmethoxy)propyl]-2,4-dihydro-4-[4-[4-(4-hydroxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one, TRC
CAS: 184177-83-1 Molecular Formula: C30 H35 N5 O3 Molecular Weight (g/mol): 513.63 Synonym: [S-(R*,R*)]-2-[1-Ethyl-2-(phenylmethoxy)propyl]-2,4-dihydro-4-[4-[4-(4-hydroxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one,Posaconazole Impurity IUPAC Name: 4-[4-[4-(4-hydroxyphenyl)piperazin-1-yl]phenyl]-2-[(2S,3S)-2-phenylmethoxypentan-3-yl]-1,2,4-triazol-3-one SMILES: CC[C@@H]([C@H](C)OCc1ccccc1)N2N=CN(C2=O)c3ccc(cc3)N4CCN(CC4)c5ccc(O)cc5