missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress TRHR Total GPCR Internalization Assay
Description
The PathHunter™ TRHR Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. TRH) based internalization of TRHR via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged TRHR and a PK-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the TRHR receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Total Internalization |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | TRH |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | TRHR |
| Cell Line | U2OS |
| Target | TRHR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?