missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress TORC2 Nuclear Translocation Assay
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ eXpress TORC2 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the CRTC/CREB pathway. This kit assesses ligand (e.g. forskolin) based pathway activation via detection of resulting TORC2 (CRTC2) nuclear translocation. This product uses EFC technology. The included cells overexpress PL-tagged TORC2 (CRTC2) and nuclear localized EA. Upon pathway activation, induced nuclear translocation of TORC2 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Pathway |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | TORC2 (CRTC2) |
| Target | TORC2 (CRTC2) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction