missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress NMBR CHO-K1 β-Arrestin GPCR Assay

Product Code. 17334019
Encompass
Change view
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17334019 1,000 dp (10 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17334019 Supplier DiscoverX Supplier No. 930925E2CP2L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress NMBR CHO-K1 β-Arrestin GPCR Assay measures NMBR (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ NMBR β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. neuromedin B) based activation of NMBR GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged NMBR and EA-tagged β-Arrestin 2. Activation of NMBR stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry β-Arrestin-2 recruitment
For Use With (Application) Assay-Ready eXpress Kits
Primary Agonist Neuromedin B (porcine)
Quantity 1,000 dp (10 x 96-well)
Target Entry NMBR
Cell Line CHO-K1
Target NMBR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.