missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress Keap1-NRF2 Nuclear Translocation Assay
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ eXpress Keap1-NRF2 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the Keap1-NRF2 pathway. This kit assesses ligand (e.g. methyl CDDO) based pathway activation via detection of resulting NRF2 nuclear translocation. This product uses EFC technology. The included cells overexpress PL-tagged NRF2 and nuclear localized EA alongside untagged Keap1. Upon pathway activation, induced nuclear translocation of NRF2 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Pathway |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | Keap1-NRF2 |
| Target | Keap1-NRF2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction