missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress HEK 293 RELA Nuclear Translocation Assay
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ eXpress HEK 293 RELA Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the NFκB pathway. This kit assesses ligand (e.g. wortmannin) based pathway activation via detection of resulting RELA nuclear translocation. This product uses EFC technology. The included cells overexpress PL-tagged RELA and nuclear localized EA. Upon pathway activation, induced nuclear translocation of RELA leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Translocation |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | IL-1α |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | NFκB |
| Cell Line | 293H |
| Target | NFκB |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction