missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress AVPR1B Activated GPCR Internalization Assay

Product Code. 17333719
Encompass
Change view
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Product Code. Quantity unitSize
17333719 200 dp (2 x 96-well) 1 set
1 options
This item is not returnable. View return policy

Product Code. 17333719

Brand: DiscoverX 930722E3CP5M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress AVPR1B Activated GPCR Internalization Assay measures AVPR1B (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ AVPR1B Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. vasopressin) based internalization of AVPR1B via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged AVPR1B. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting AVPR1B-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry GPCR
For Use With (Application) Assay-Ready eXpress Kits
Quantity 200 dp (2 x 96-well)
Target Entry AVPR1B
Target AVPR1B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.