missing translation for 'onlineSavingsMsg'
Learn More

(1S)-10-Camphorsulfonamide, 97%

Product Code. 11448520
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448520 25 g 25g
11458520 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448520 Supplier Thermo Scientific Alfa Aesar Supplier No. 044361.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 60933-63-3 | C10H17NO3S | 231.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60933-63-3
Molecular Formula C10H17NO3S
Molecular Weight (g/mol) 231.31
MDL Number MFCD00075611,MFCD00075611,MFCD00151500
InChI Key SBLUNABTQYDFJM-GMSGAONNSA-N
Synonym 1s-10-camphorsulfonamide, 1r,4s-7,7-dimethyl-3-oxo-4-bicyclo 2.2.1 heptanyl methanesulfonamide, 1r---10-camphorsulfonamide, 1s-+-10-camphorsulfonamide, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonamide, bicyclo 2.2.1 heptane-1-methanesulfonamide,7,7-dimethyl-2-oxo-, 1s,4r, 1r,4s-7,7-dimethyl-3-oxidanylidene-4-bicyclo 2.2.1 heptanyl methanesulfonamide, 1-1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonamide
PubChem CID 12840037
IUPAC Name [(4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonamide
SMILES CC1(C)[C@@H]2CC[C@@]1(CS(N)(=O)=O)C(=O)C2

Spécification

Melting Point 133°C to 135°C (decomposition)
Quantity 25 g
Formula Weight 231.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (1S)-10-Camphorsulfonamide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.