accessibility menu, dialog, popup

UserName

CAS RN 60933-63-3

H3CH3C(R)(S)SH2NOOO

CAS RN 60933-63-3

IUPAC Name: [(1S,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonamide
Synonyms: [(1S,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonamide
Molecular Weight (g/mol): 231.31
Molecular Formula: C10H17NO3S
InChi Key: SBLUNABTQYDFJM-GMSGAONNSA-N
SMILES: CC1(C)[C@@H]2CC[C@@]1(CS(N)(=O)=O)C(=O)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results