missing translation for 'onlineSavingsMsg'
Learn More

(1S)-10-Camphorsulfonamide, 97%, Thermo Scientific Chemicals

Catalog Number 11458520
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
60933-63-3
C10H17NO3S
231.31
MFCD00075611,MFCD00075611,MFCD00151500
SBLUNABTQYDFJM-GMSGAONNSA-N
1s-10-camphorsulfonamide, 1r,4s-7,7-dimethyl-3-oxo-4-bicyclo 2.2.1 heptanyl methanesulfonamide, 1r---10-camphorsulfonamide, 1s-+-10-camphorsulfonamide, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonamide, bicyclo 2.2.1 heptane-1-methanesulfonamide,7,7-dimethyl-2-oxo-, 1s,4r, 1r,4s-7,7-dimethyl-3-oxidanylidene-4-bicyclo 2.2.1 heptanyl methanesulfonamide, 1-1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonamide
12840037
[(4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonamide
CC1(C)[C@@H]2CC[C@@]1(CS(N)(=O)=O)C(=O)C2
This item is not returnable. View return policy
60933-63-3
C10H17NO3S
231.31
MFCD00075611,MFCD00075611,MFCD00151500
SBLUNABTQYDFJM-GMSGAONNSA-N
1s-10-camphorsulfonamide, 1r,4s-7,7-dimethyl-3-oxo-4-bicyclo 2.2.1 heptanyl methanesulfonamide, 1r---10-camphorsulfonamide, 1s-+-10-camphorsulfonamide, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonamide, bicyclo 2.2.1 heptane-1-methanesulfonamide,7,7-dimethyl-2-oxo-, 1s,4r, 1r,4s-7,7-dimethyl-3-oxidanylidene-4-bicyclo 2.2.1 heptanyl methanesulfonamide, 1-1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonamide
12840037
[(4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonamide
CC1(C)[C@@H]2CC[C@@]1(CS(N)(=O)=O)C(=O)C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 133°C to 135°C (decomposition)
Quantity 5 g
Formula Weight 231.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (1S)-10-Camphorsulfonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'