missing translation for 'onlineSavingsMsg'
Learn More

(1R,4S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-en-1-ol, TRC

Product Code. 30474517
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474517 5 g 5g
30479660 1 g 1g
30467395 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474517 Supplier Toronto Research Chemicals Supplier No. TRCM3383405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82769-01-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.23
Synonym (1R,4S)-1-Methyl-4-(1-methylethenyl)-2-cyclohexen-1-ol, (-)-(1R,4S)-p-Mentha-2,8-dien-1-ol, (-)-p-Mentha-2,8-dien-1-ol
IUPAC Name (1R,4S)-1-methyl-4-prop-1-en-2-ylcyclohex-2-en-1-ol
SMILES O[C@@]1(C)C=C[C@@H](C(C)=C)CC1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H16O/c1-8(2)9-4-6-10(3,11)7-5-9/h4,6,9,11H,1,5,7H2,2-3H3/t9-,10+/m1/s1
Formula Weight 152.1201
Chemical Name or Material (1R,4S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-en-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.