CAS RN 82769-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R,4S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-en-1-ol, TRC
CAS: 82769-01-5 Molecular Formula: C10H16O Molecular Weight (g/mol): 152.23 Synonym: (1R,4S)-1-Methyl-4-(1-methylethenyl)-2-cyclohexen-1-ol,(-)-(1R,4S)-p-Mentha-2,8-dien-1-ol,(-)-p-Mentha-2,8-dien-1-ol IUPAC Name: (1R,4S)-1-methyl-4-prop-1-en-2-ylcyclohex-2-en-1-ol SMILES: O[C@@]1(C)C=C[C@@H](C(C)=C)CC1